missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 179.14 |
Percent Purity | 97% |
Chemical Name or Material | [5-Methyl-2-(trifluoromethyl)-3-furyl]methylamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'