missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2-(trifluoromethyl)-3-furyl isocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 191.11 |
Percent Purity | 90% |
Chemical Name or Material | 5-Methyl-2-(trifluoromethyl)-3-furyl isocyanate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'