missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-4509801
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
306935-02-4
C6H4ClF3O3S
248.6
GSYXWDYWBRICSD-UHFFFAOYSA-N
5-methyl-2-trifluoromethyl furan-3-sulfonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonyl chloride, 3-furansulfonylchloride, 5-methyl-2-trifluoromethyl, 5-methyl-2-trifluoromethyl furan-3-sulphonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonylchloride, 5-methyl-2-trifluoromethyl-furan-3-sulfonyl chloride, chloro 5-methyl-2-trifluoromethyl 3-furyl sulfone, 3-furansulfonyl chloride, 5-methyl-2-trifluoromethyl-9ci
2779895
5-methyl-2-(trifluoromethyl)furan-3-sulfonyl chloride
CC1=CC(=C(O1)C(F)(F)F)S(=O)(=O)Cl
This item is not returnable. View return policy
306935-02-4
C6H4ClF3O3S
248.6
GSYXWDYWBRICSD-UHFFFAOYSA-N
5-methyl-2-trifluoromethyl furan-3-sulfonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonyl chloride, 3-furansulfonylchloride, 5-methyl-2-trifluoromethyl, 5-methyl-2-trifluoromethyl furan-3-sulphonyl chloride, 5-methyl-2-trifluoromethyl-3-furansulfonylchloride, 5-methyl-2-trifluoromethyl-furan-3-sulfonyl chloride, chloro 5-methyl-2-trifluoromethyl 3-furyl sulfone, 3-furansulfonyl chloride, 5-methyl-2-trifluoromethyl-9ci
2779895
5-methyl-2-(trifluoromethyl)furan-3-sulfonyl chloride
CC1=CC(=C(O1)C(F)(F)F)S(=O)(=O)Cl
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furansulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'