missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methyl-2-phenylbenzimidazole, 95%

Brand:  Alfa Aesar™ H64405.06

Code : NEW

Additional Details : CAS Number : 2963-65-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15401609

  • £478.06 / 5g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

5-Methyl-2-phenylbenzimidazole
2963-65-7
208.26
MFCD00458486

Specifications

5-Methyl-2-phenylbenzimidazole
2963-65-7
95%
MFCD00458486
5g
Solid
208.26
Videos
SDS