missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-methyl-2-phenyl-2H-1,2,3-triazole-4-carbonyl chloride, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 221.65 |
Percent Purity | 97% |
Chemical Name or Material | 5-Methyl-2-phenyl-2H-1, 2, 3-triazole-4-carbonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'