missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-4510312
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
18735-74-5
C11H8NO3
202.19
MFCD00275475
YABCPNYCFFUVNM-UHFFFAOYSA-M
5-methyl-2-phenyloxazole-4-carboxylic acid, 2-phenyl-5-methyloxazole-4-carboxylic acid, 5-methyl-2-phenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazole-4-carboxylicacid, 4-carboxy-5-methyl-2-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid,5-methyl-2-phenyl, 2-phenyl-5-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylicacid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazol-4-ylcarboxylic acid
318533
5-methyl-2-phenyl-1,3-oxazole-4-carboxylic acid
CC1=C(N=C(O1)C1=CC=CC=C1)C([O-])=O
This item is not returnable. View return policy
18735-74-5
C11H8NO3
202.19
MFCD00275475
YABCPNYCFFUVNM-UHFFFAOYSA-M
5-methyl-2-phenyloxazole-4-carboxylic acid, 2-phenyl-5-methyloxazole-4-carboxylic acid, 5-methyl-2-phenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazole-4-carboxylicacid, 4-carboxy-5-methyl-2-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid,5-methyl-2-phenyl, 2-phenyl-5-methyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylicacid, 5-methyl-2-phenyl, 5-methyl-2-phenyloxazol-4-ylcarboxylic acid
318533
5-methyl-2-phenyl-1,3-oxazole-4-carboxylic acid
CC1=C(N=C(O1)C1=CC=CC=C1)C([O-])=O
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 203.2
Percent Purity 97%
Chemical Name or Material 5-Methyl-2-phenyl-1, 3-oxazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'