missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-nitrobenzoic acid, 95%, ACROS Organics™

Brand:  Acros Organics 151090050

Code : 29

Additional Details : CAS Number : 3113-72-2 Weight : 0.05500kg


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 11904991

  • £33.60 / 5g


EU Stock 3
Add to basket


Specifications

Chemical Identifiers

5-Methyl-2-nitrobenzoic acid
94.0
95%
C8H7NO4
181.15
3113-72-2
100.0
95%
CH3C6H3(NO2)CO2H
6-Nitro-m-toluic acid

Specifications

5-Methyl-2-nitrobenzoic acid
94.0
Authentic
Powder or Crystalline Powder
95%
CH3C6H3(NO2)CO2H
White to yellow
133.0° to 136.0°C
3113-72-2
100.0
Glass bottle
95%
C8H7NO4
181.15
6-Nitro-m-toluic acid
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Methyl-2-nitrobenzoic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS