missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-nitrobenzoic acid, 95%, Thermo Scientific™

Catalog Number p-4229304
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3113-72-2
C8H7NO4
181.15
QRRSIFNWHCKMSW-UHFFFAOYSA-N
5-methyl-2-nitrobenzoic acid, 2-nitro-5-methylbenzoic acid, benzoic acid, 5-methyl-2-nitro, 3-methyl-6-nitrobenzoic acid, 6-nitro-m-toluic acid, m-toluic acid, 6-nitro, 6-nitro-p-toluic acid, 5-methyl-2-nitrobenzoicacid, 5-methyl-2-nitro-benzoic acid, benzoic acid,5-methyl-2-nitro
18371
CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O
This item is not returnable. View return policy
3113-72-2
C8H7NO4
181.15
QRRSIFNWHCKMSW-UHFFFAOYSA-N
5-methyl-2-nitrobenzoic acid, 2-nitro-5-methylbenzoic acid, benzoic acid, 5-methyl-2-nitro, 3-methyl-6-nitrobenzoic acid, 6-nitro-m-toluic acid, m-toluic acid, 6-nitro, 6-nitro-p-toluic acid, 5-methyl-2-nitrobenzoicacid, 5-methyl-2-nitro-benzoic acid, benzoic acid,5-methyl-2-nitro
18371
CC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O
CAS Min % 94.0
CAS Max % 100.0
Melting Point 133.0°C to 136.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)CO2H
Quantity 5 g
Beilstein 09,482
Formula Weight 181.15
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 5-Methyl-2-nitrobenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methyl-2-nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'