missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-nitroaniline, 95%, Acros Organics

Brand:  Acros Organics 441620010

UNSPSC : 12161500

Code : 29

Additional Details : CAS Number : 578-46-1 Weight : 0.05100kg Transport : UN number : 2660 Chem class : 1 Pack group : III


Quantity 1g

 View more versions of this product

Product Code. 11965221

  • £15.00 / 1g


Estimated Shipment
13-09-2017
Add to basket


Specifications

Chemical Identifiers

5-Methyl-2-nitroaniline
95%
C7H8N2O2
152.15

Specifications

5-Methyl-2-nitroaniline
95%
C7H8N2O2
104.5° to 105.5°C
578-46-1
95%
152.15
1g
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Methyl-2-nitroaniline

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

Documentation

Technical Documents