missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furanmethanamine, 98%, Thermo Scientific Chemicals

Catalog Number p-163179
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14003-16-8
C6H9NO
111.14
MFCD00143471
YSEAGSCGERFGBL-UHFFFAOYSA-N
5-methylfurfurylamine, 5-methylfuran-2-yl methanamine, 5-methyl-2-furanmethanamine, 2-aminomethyl-5-methylfuran, 5-methyl-2-furyl methylamine, 2-furanmethanamine, 5-methyl, 5-methyl-2-furyl methyl amine, 1-5-methylfuran-2-yl methanamine, pubchem6995, acmc-209cjz
2724683
(5-methylfuran-2-yl)methanamine
CC1=CC=C(CN)O1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
14003-16-8
C6H9NO
111.14
MFCD00143471
YSEAGSCGERFGBL-UHFFFAOYSA-N
5-methylfurfurylamine, 5-methylfuran-2-yl methanamine, 5-methyl-2-furanmethanamine, 2-aminomethyl-5-methylfuran, 5-methyl-2-furyl methylamine, 2-furanmethanamine, 5-methyl, 5-methyl-2-furyl methyl amine, 1-5-methylfuran-2-yl methanamine, pubchem6995, acmc-209cjz
2724683
(5-methylfuran-2-yl)methanamine
CC1=CC=C(CN)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -22°C
Color Yellow
Density 1.0100g/mL
Boiling Point 57°C (13.0 Mbar)
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4860 to 1.488
Quantity 5 g
Specific Gravity 1.01
Solubility Information Solubility in water: >1000g/L (20°C)
Formula Weight 111.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-furanmethanamine, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 5-Methyl-2-furanmethanamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'