missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methyl-2-(5-methyl-2-furyl)benzimidazole, 95%

Brand:  Alfa Aesar™ H64625.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15451909

  • £612.68 / 5g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Methyl-2-(5-methyl-2-furyl)benzimidazole
212.25
MFCD21642610

Specifications

5-Methyl-2-(5-methyl-2-furyl)benzimidazole
95%
MFCD21642610
Solid
212.25
5g
Videos
SDS