missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methyl-2-[4-(methylthio)phenyl]benzimidazole, 95%

Brand:  Alfa Aesar™ H64827.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459176

  • £648.72 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Methyl-2-[4-(methylthio)phenyl]benzimidazole
254.35
MFCD15173048

Specifications

5-Methyl-2-[4-(methylthio)phenyl]benzimidazole
95%
MFCD15173048
Solid
254.35
5g
Videos
SDS