missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-(4-methylphenyl)benzimidazole, 95%, Thermo Scientific™

Catalog Number 15452319
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
7118-66-3
C15H14N2
222.291
MFCD00436743
OPBSAVHQPYKEQP-UHFFFAOYSA-N
5-methyl-2-p-tolyl-1h-benzoimidazole, 5-methyl-2-4-methylphenyl-1h-benzimidazole, 6-methyl-2-4-methylphenyl-1h-benzimidazole, 5-methyl-2-4-methylphenyl benzimidazole, 5-methyl-2-4-methylphenyl-1h-1,3-benzodiazole, 5-methyl-2-4-methylphenyl-3h-1,3-benzodiazole, 2-4-methylphenyl-5-methyl-1h-benzimidazole, 1h-benzimidazole, 5-methyl-2-4-methylphenyl
893072
6-methyl-2-(4-methylphenyl)-1H-benzimidazole
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C
This item is not returnable. View return policy
7118-66-3
C15H14N2
222.291
MFCD00436743
OPBSAVHQPYKEQP-UHFFFAOYSA-N
5-methyl-2-p-tolyl-1h-benzoimidazole, 5-methyl-2-4-methylphenyl-1h-benzimidazole, 6-methyl-2-4-methylphenyl-1h-benzimidazole, 5-methyl-2-4-methylphenyl benzimidazole, 5-methyl-2-4-methylphenyl-1h-1,3-benzodiazole, 5-methyl-2-4-methylphenyl-3h-1,3-benzodiazole, 2-4-methylphenyl-5-methyl-1h-benzimidazole, 1h-benzimidazole, 5-methyl-2-4-methylphenyl
893072
6-methyl-2-(4-methylphenyl)-1H-benzimidazole
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 222.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5-Methyl-2-(4-methylphenyl)benzimidazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'