missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-1H-tetrazole, 97%, Thermo Scientific Chemicals
102.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.
Solubility
Soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.
Solubility
Soluble in water.
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
Melting Point | 142°C to 146°C |
Quantity | 25 g |
Solubility Information | Soluble in water. |
Formula Weight | 84.08 |
Percent Purity | 97% |
Chemical Name or Material | 5-Methyl-1H-tetrazole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'