missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-benzotriazole, 98%, Thermo Scientific™

Catalog Number p-160320
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
136-85-6
C7H7N3
133.15
MFCD00005702
LRUDIIUSNGCQKF-UHFFFAOYSA-N
5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1
8705
CHEBI:83455
5-methyl-2H-benzotriazole
CC1=CC2=NNN=C2C=C1
This item is not returnable. View return policy
136-85-6
C7H7N3
133.15
MFCD00005702
LRUDIIUSNGCQKF-UHFFFAOYSA-N
5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1
8705
CHEBI:83455
5-methyl-2H-benzotriazole
CC1=CC2=NNN=C2C=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 80.0°C to 84.0°C
Color Cream to Beige
Boiling Point 210.0°C to 212.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 26, 58
Solubility Information Solubility in water: 6.0g/L (25 c). Other solubilities: soluble in acids and alkalies,soluble in methanol and ethanol
Formula Weight 133.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-1H-benzotriazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methyl-1H-benzotriazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'