missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080879
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
136-85-6
C7H7N3
133.15
MFCD00005702
LRUDIIUSNGCQKF-UHFFFAOYSA-N
5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1
8705
CHEBI:83455
5-methyl-2H-benzotriazole
CC1=CC2=NNN=C2C=C1
This item is not returnable. View return policy
136-85-6
C7H7N3
133.15
MFCD00005702
LRUDIIUSNGCQKF-UHFFFAOYSA-N
5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1
8705
CHEBI:83455
5-methyl-2H-benzotriazole
CC1=CC2=NNN=C2C=C1

5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

Solubility
Soluble in chloroform, methanol, water (6.0 g/L at 25°C), ethanol, and toluene (slightly).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 77°C to 84°C
pH 5.3
Boiling Point 210°C to 212°C (12 mmHg)
Odor Odorless
Quantity 25 g
Beilstein 116658
Solubility Information Soluble in chloroform,methanol,water (6.0g/L at 25°C),ethanol,and toluene (slightly).
Formula Weight 133.15
Percent Purity ≥98%
Chemical Name or Material 5-Methyl-1H-benzotriazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'