missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxyresorcinol, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
5-Methoxyresorcinol was used in synthesis of isorobustin, substituted linear and angular benzofurocoumarins.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 78°C to 80°C |
Color | Yellow |
Boiling Point | 188°C to 189°C (12 mmHg) |
Quantity | 250 mg |
Beilstein | 1423578 |
Solubility Information | Soluble in water. |
Formula Weight | 140.14 |
Percent Purity | 95% |
Physical Form | Solid |
Chemical Name or Material | 5-Methoxyresorcinol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'