missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole, 99%, Thermo Scientific Chemicals

Catalog Number 10449240
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1006-94-6
C9H9NO
147.18
MFCD00005674
DWAQDRSOVMLGRQ-UHFFFAOYSA-N
5-methoxyindole, 1h-indole, 5-methoxy, femedol, 5-methoxy indole, indol-5-yl methyl ether, indole, 5-methoxy, methoxy-5 indole, unii-dqm3as43pq, methoxy-5 indole french, methoxyindole 5-
13872
5-methoxy-1H-indole
COC1=CC2=C(C=C1)NC=C2
This item is not returnable. View return policy
1006-94-6
C9H9NO
147.18
MFCD00005674
DWAQDRSOVMLGRQ-UHFFFAOYSA-N
5-methoxyindole, 1h-indole, 5-methoxy, femedol, 5-methoxy indole, indol-5-yl methyl ether, indole, 5-methoxy, methoxy-5 indole, unii-dqm3as43pq, methoxy-5 indole french, methoxyindole 5-
13872
5-methoxy-1H-indole
COC1=CC2=C(C=C1)NC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 52.0°C to 56.0°C
Color Brown to White
Boiling Point 176.0°C to 178.0°C (17.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, II, 44
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone
Formula Weight 147.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyindole, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxyindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'