missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole, 99%, Thermo Scientific Chemicals

Catalog Number p-7028144
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1006-94-6
C9H9NO
147.177
MFCD00005674
DWAQDRSOVMLGRQ-UHFFFAOYSA-N
5-methoxyindole, 1h-indole, 5-methoxy, femedol, 5-methoxy indole, indol-5-yl methyl ether, indole, 5-methoxy, methoxy-5 indole, unii-dqm3as43pq, methoxy-5 indole french, methoxyindole 5-
13872
5-methoxy-1H-indole
COC1=CC2=C(C=C1)NC=C2
This item is not returnable. View return policy
1006-94-6
C9H9NO
147.177
MFCD00005674
DWAQDRSOVMLGRQ-UHFFFAOYSA-N
5-methoxyindole, 1h-indole, 5-methoxy, femedol, 5-methoxy indole, indol-5-yl methyl ether, indole, 5-methoxy, methoxy-5 indole, unii-dqm3as43pq, methoxy-5 indole french, methoxyindole 5-
13872
5-methoxy-1H-indole
COC1=CC2=C(C=C1)NC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 52°C to 55°C
Boiling Point 176°C to 178°C (17 mmHg)
Quantity 1 g
Beilstein 116722
Sensitivity Light sensitive
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 5-Methoxyindole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'