missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7055992
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
10242-01-0
C10H9NO3
191.186
MFCD03265451
RVVSEZGJCOAUED-UHFFFAOYSA-N
5-methoxyindole-3-carboxylic acid, 5-methoxy-3-indolecarboxylic acid, 5-methoxy-3-carboxyindole, 1h-indole-3-carboxylic acid, 5-methoxy, pubchem7984, zlchem 1276, 1h-indole-3-carboxylicacid, 5-methoxy, 5-methoxyindole-3-carboxylic acid;, 5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
259191
5-methoxy-1H-indole-3-carboxylic acid
COC1=CC2=C(C=C1)NC=C2C(=O)O
This item is not returnable. View return policy
10242-01-0
C10H9NO3
191.186
MFCD03265451
RVVSEZGJCOAUED-UHFFFAOYSA-N
5-methoxyindole-3-carboxylic acid, 5-methoxy-3-indolecarboxylic acid, 5-methoxy-3-carboxyindole, 1h-indole-3-carboxylic acid, 5-methoxy, pubchem7984, zlchem 1276, 1h-indole-3-carboxylicacid, 5-methoxy, 5-methoxyindole-3-carboxylic acid;, 5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
259191
5-methoxy-1H-indole-3-carboxylic acid
COC1=CC2=C(C=C1)NC=C2C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Red
Quantity 250 mg
Beilstein 475711
Formula Weight 191.19
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material 5-Methoxyindole-3-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'