missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-3-carboxaldehyde, 99+%, Thermo Scientific™

Catalog Number 10595881
Click to view available options
Quantity:
100 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10601-19-1
C10H9NO2
175.19
MFCD00005623
TUWARWGEOHQXCO-UHFFFAOYSA-N
5-methoxyindole-3-carboxaldehyde, 3-formyl-5-methoxyindole, 5-methoxyindole-3-carboxyaldehyde, 5-methoxyindole-3-carbaldehyde, 5-methoxyindole-3-aldehyde, 5-methoxy-3-formylindole, 1h-indole-3-carboxaldehyde, 5-methoxy, 5-methoxy-indol-3-aldehyde, unii-63c50zw2d0, 5-methoxy-1h-indole-3-carboxaldehyde
82758
5-methoxy-1H-indole-3-carbaldehyde
COC1=CC=C2NC=C(C=O)C2=C1
This item is not returnable. View return policy
10601-19-1
C10H9NO2
175.19
MFCD00005623
TUWARWGEOHQXCO-UHFFFAOYSA-N
5-methoxyindole-3-carboxaldehyde, 3-formyl-5-methoxyindole, 5-methoxyindole-3-carboxyaldehyde, 5-methoxyindole-3-carbaldehyde, 5-methoxyindole-3-aldehyde, 5-methoxy-3-formylindole, 1h-indole-3-carboxaldehyde, 5-methoxy, 5-methoxy-indol-3-aldehyde, unii-63c50zw2d0, 5-methoxy-1h-indole-3-carboxaldehyde
82758
5-methoxy-1H-indole-3-carbaldehyde
COC1=CC=C2NC=C(C=O)C2=C1
CAS Min % 99.0
CAS Max % 100.0
Melting Point 180.0°C to 184.0°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, II, 424
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 175.19
Percent Purity 99+%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 5-Methoxyindole-3-carboxaldehyde, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxyindole-3-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'