missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxyindole-2-carboxylic acid, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | 197.0°C to 201.0°C |
Color | Brown to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 10 g |
Beilstein | 22, II, 169 |
Solubility Information | Solubility in water: slightly soluble in water. Other solubilities: soluble in acetone |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'