missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Methoxy-3-(4-methyl-1-piperazinylmethyl)indole, 95%

Brand:  Alfa Aesar™ H63056.03

Code : NEW

Additional Details : CAS Number : 701205-15-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448106

  • £95.61 / 1g


Estimated Shipment date
from Supplier 08-03-2018
Add to basket

Specifications

Chemical Identifiers

5-Methoxy-3-(4-methyl-1-piperazinylmethyl)indole
701205-15-4
259.35
11950927

Specifications

5-Methoxy-3-(4-methyl-1-piperazinylmethyl)indole
701205-15-4
95%
11950927
1g
Solid
259.35
Videos
SDS