missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylaniline, 97%, Thermo Scientific™

Catalog Number 10693225
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
50868-72-9
C8H11NO
137.18
MFCD00075057
RPJXLEZOFUNGNZ-UHFFFAOYSA-N
5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic
99500
5-methoxy-2-methylaniline
COC1=CC=C(C)C(N)=C1
This item is not returnable. View return policy
50868-72-9
C8H11NO
137.18
MFCD00075057
RPJXLEZOFUNGNZ-UHFFFAOYSA-N
5-methoxy-o-toluidine, 2-amino-4-methoxytoluene, benzenamine, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzenamine, 2-methyl-5-ethoxyaniline, 5-methoxy-2-methylphenylamine, 6-methyl-m-anisidine, 3-amino-4-methylanisole 5-methoxy-2-methylaniline, pubchem19827, acmc-1awic
99500
5-methoxy-2-methylaniline
COC1=CC=C(C)C(N)=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 43°C to 46°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H3(CH3)NH2
Quantity 1 g
Formula Weight 137.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 5-Methoxy-2-methylaniline
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxy-2-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'