missing translation for 'onlineSavingsMsg'
Learn More

5-Isoxazol-5-ylthiophene-2-sulfonyl chloride, Tech., Thermo Scientific™

Catalog Number p-6830004
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
551930-53-1
C7H4ClNO3S2
249.68
SSGKBJJYLTYNQD-UHFFFAOYSA-N
5-5-isoxazyl thiophene-2-sulfonyl chloride, 5-isoxazol-5-ylthiophene-2-sulfonyl chloride, 5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonyl chloride, 5-isoxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-isoxazol-5-yl 2-thienyl sulfone, ssgkbjjyltynqd-uhfffaoysa-n, 5-5-isoxazyl thiophene-2-sulphonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonoyl chloride, 5-5-isoxazolyl thiophene-2-sulfonyl chloride
2795330
This item is not returnable. View return policy
551930-53-1
C7H4ClNO3S2
249.68
SSGKBJJYLTYNQD-UHFFFAOYSA-N
5-5-isoxazyl thiophene-2-sulfonyl chloride, 5-isoxazol-5-ylthiophene-2-sulfonyl chloride, 5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonyl chloride, 5-isoxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-isoxazol-5-yl 2-thienyl sulfone, ssgkbjjyltynqd-uhfffaoysa-n, 5-5-isoxazyl thiophene-2-sulphonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonoyl chloride, 5-5-isoxazolyl thiophene-2-sulfonyl chloride
2795330
Quantity 250 mg
Formula Weight 249.7
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-Isoxazol-5-ylthiophene-2-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'