missing translation for 'onlineSavingsMsg'
Learn More

5-(Isocyanatomethyl)-1,3-benzodioxole, ≥97%, Thermo Scientific™

Catalog Number p-3175433
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
71217-46-4
C9H7NO3
177.159
MFCD08435909
RIUNOJGBBOBVDE-UHFFFAOYSA-N
5-isocyanatomethyl-2h-1,3-benzodioxole, 5-isocyanatomethyl-1,3-benzodioxole, 1,3-benzodioxole,5-isocyanatomethyl, 1,3-benzodioxole, 5-isocyanatomethyl, 3,4-methylenedioxy benzyl isocyanate, 1,3-benzodioxol-5-ylmethyl isocyanate, 5-isocyanatomethyl benzo d 1,3 dioxole, 2h-benzo 3,4-d 1,3-dioxolan-5-ylmethanisocyanate
15932354
5-(isocyanatomethyl)-1,3-benzodioxole
C1OC2=C(O1)C=C(C=C2)CN=C=O
This item is not returnable. View return policy
71217-46-4
C9H7NO3
177.159
MFCD08435909
RIUNOJGBBOBVDE-UHFFFAOYSA-N
5-isocyanatomethyl-2h-1,3-benzodioxole, 5-isocyanatomethyl-1,3-benzodioxole, 1,3-benzodioxole,5-isocyanatomethyl, 1,3-benzodioxole, 5-isocyanatomethyl, 3,4-methylenedioxy benzyl isocyanate, 1,3-benzodioxol-5-ylmethyl isocyanate, 5-isocyanatomethyl benzo d 1,3 dioxole, 2h-benzo 3,4-d 1,3-dioxolan-5-ylmethanisocyanate
15932354
5-(isocyanatomethyl)-1,3-benzodioxole
C1OC2=C(O1)C=C(C=C2)CN=C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-(Isocyanatomethyl)-1, 3-benzodioxole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'