missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 5-isocyanato-1-methyl-1h-indole, 1h-indole, 5-isocyanato-1-methyl, 1-methylindol-5-isocyanate, 1h-indole,5-isocyanato-1-methyl, 1-methyl-1h-indole-5-yl isocyanate |
IUPAC Name | 5-isocyanato-1-methylindole |
Formula Weight | 172.19 |
Percent Purity | 97% |
Chemical Name or Material | 5-Isocyanato-1-methyl-1H-indole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'