missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-methylaniline, 97%, Thermo Scientific™

Catalog Number p-4813226
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
83863-33-6
C7H8IN
233.05
MFCD00051531
IOEHXNCBPIBDBZ-UHFFFAOYSA-N
2-amino-4-iodotoluene, 5-iodo-o-toluidine, 5-iodo-2-methyl-phenylamine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl, 5-iodo-2-methyl-aniline, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 2-methyl-5-iodoaniline, #
522802
5-iodo-2-methylaniline
CC1=CC=C(I)C=C1N
This item is not returnable. View return policy
83863-33-6
C7H8IN
233.05
MFCD00051531
IOEHXNCBPIBDBZ-UHFFFAOYSA-N
2-amino-4-iodotoluene, 5-iodo-o-toluidine, 5-iodo-2-methyl-phenylamine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl, 5-iodo-2-methyl-aniline, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 2-methyl-5-iodoaniline, #
522802
5-iodo-2-methylaniline
CC1=CC=C(I)C=C1N
Melting Point 48°C to 50°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 233.05
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-methylaniline
compliance-icons compliance-icons
Product Identifier
  • 5-Iodo-2-methylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'