missing translation for 'onlineSavingsMsg'
Learn More

(+)-5-Iodo-2'-deoxyuridine, 99%, Thermo Scientific Chemicals

Catalog Number p-3736739
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
54-42-2
C9H11IN2O5
354.10
MFCD00134656
XQFRJNBWHJMXHO-RRKCRQDMSA-N
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
5905
CHEBI:147675
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O
This item is not returnable. View return policy
54-42-2
C9H11IN2O5
354.10
MFCD00134656
XQFRJNBWHJMXHO-RRKCRQDMSA-N
idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
5905
CHEBI:147675
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100
Melting Point 155.0°C to 180.0°C
Color Beige to White
Loss on Drying 1% max. (1 g, 60°C) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4931
Specific Rotation Condition + 280.00 (20.00°C c=1,1M NaOH)
Specific Rotation + 280.00
Solubility Information Solubility in water: 1.6g/L (20°C). Other solubilities: 2.0g/L hcl 0.2n- 74.0g/L naoh 0.2n (25°C),4.4g/L methanol- 2.6g/L alcohol (25°C),0.014g/L ether- 0.003g/L chloroform (25°C),1.6g/L acetone- 1.8g/L ethyl acetate (25°C),5.7
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
Formula Weight 354.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (+)-5-Iodo-2′-deoxyuridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (+)-5-Iodo-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'