missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-deoxycytidine, 99%, Thermo Scientific Chemicals

Catalog Number p-7042101
Click to view available options
Quantity:
1 g
2 g
This item is not returnable. View return policy
611-53-0
C9H12IN3O4
353.12
MFCD00038063
WEVJJMPVVFNAHZ-RRKCRQDMSA-N
5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
65050
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one
NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1
This item is not returnable. View return policy
611-53-0
C9H12IN3O4
353.12
MFCD00038063
WEVJJMPVVFNAHZ-RRKCRQDMSA-N
5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
65050
4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidin-2-one
NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1

5-Iodo-2'-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2?-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4,5:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-2′-deoxycytidine is used in the construction of DNA oligomers to enable structural studies and photoactivated cross-linking. It is used in the synthesis of other modified nucleosides, such as 5-ethynylferrocenyl-2′-deoxycytidine used in semiconductor electrodes and 10-(2-deoxyβ-D-ribofuranosyl)pyrimido[4′,5′:4,5]pyrimido[1,6-a]indole-6,9(7H)-dione (dCPPI).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Melting Point 180°C to 183°C
Color White
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 353.11
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Iodo-2'-deoxycytidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'