missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-1H-indazole, 95%, Thermo Scientific Chemicals

Catalog Number 10545680
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
55919-82-9
C7H5IN2
244.03
MFCD07781642
CGCHCLICSHIAAM-UHFFFAOYSA-N
5-iodoindazole, 1h-indazole, 5-iodo, 5-iodo 1h indazole, 5-iodanyl-1h-indazole, 5-iodio-1h-indazole, pubchem20575, d07ovy, indazole, 5-iodo, 5-iodo-1h-indazole
21894739
5-iodo-1H-indazole
C1=CC2=C(C=C1I)C=NN2
This item is not returnable. View return policy
55919-82-9
C7H5IN2
244.03
MFCD07781642
CGCHCLICSHIAAM-UHFFFAOYSA-N
5-iodoindazole, 1h-indazole, 5-iodo, 5-iodo 1h indazole, 5-iodanyl-1h-indazole, 5-iodio-1h-indazole, pubchem20575, d07ovy, indazole, 5-iodo, 5-iodo-1h-indazole
21894739
5-iodo-1H-indazole
C1=CC2=C(C=C1I)C=NN2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 161°C to 163°C
Color Purple to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 244.03
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodo-1H-indazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Iodo-1H-indazole, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'