missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Hydroxypyridine-2-carboxylic acid, ≥98%

Brand:  Alfa Aesar™ H64265.06

Code : NEW

Additional Details : CAS Number : 15069-92-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15441379

  • £50.10 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Description

Description

Applications
Useful synthetic intermediate. Also intermediate for pharmaceutical application.

Solubility
Soluble in acetone, dimethyl sulfoxide , ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

5-Hydroxypyridine-2-carboxylic acid
139.11
116009
15069-92-8
MFCD00661309
5-Hydroxypicolinic acid

Specifications

5-Hydroxypyridine-2-carboxylic acid
5g
Solid
139.11
116009
260° to 264°C
15069-92-8
≥98%
MFCD00661309
5-Hydroxypicolinic acid
Videos
SDS