missing translation for 'onlineSavingsMsg'
Learn More

5-(Hydroxymethyl)-2-iodophenol,97%, Thermo Scientific™

Catalog Number 10515744
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
773869-57-1
C7H7IO2
250.035
VIWWSAGABDIIFF-UHFFFAOYSA-N
5-hydroxymethyl-2-iodophenol, 3-hydroxy-4-iodobenzyl alcohol, 3-hydroxy-4-iodobenzenemethanol, benzenemethanol,3-hydroxy-4-iodo, zlchem 794, 5-hydroxymethyl-2-iodo-phenol, 3-hydroxy-4-iodophenyl methanol, 5-hydroxymethyl-2-iodanyl-phenol
18525941
5-(hydroxymethyl)-2-iodophenol
C1=CC(=C(C=C1CO)O)I
This item is not returnable. View return policy
773869-57-1
C7H7IO2
250.035
VIWWSAGABDIIFF-UHFFFAOYSA-N
5-hydroxymethyl-2-iodophenol, 3-hydroxy-4-iodobenzyl alcohol, 3-hydroxy-4-iodobenzenemethanol, benzenemethanol,3-hydroxy-4-iodo, zlchem 794, 5-hydroxymethyl-2-iodo-phenol, 3-hydroxy-4-iodophenyl methanol, 5-hydroxymethyl-2-iodanyl-phenol
18525941
5-(hydroxymethyl)-2-iodophenol
C1=CC(=C(C=C1CO)O)I
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 250.03
Percent Purity 97%
Chemical Name or Material 5-(hydroxymethyl)-2-iodophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'