missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole, 97%, Thermo Scientific Chemicals

Catalog Number p-3736596
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1953-54-4
C8H7NO
133.15
MFCD00005677
LMIQERWZRIFWNZ-UHFFFAOYSA-N
5-hydroxyindole, indol-5-ol, 5-hydroxy-1h-indole, hydroxy-5 indole, 5-hydroxy-indole, 5-indolol, hydroxy-5-indole, 5-hydroxyindol, 5-hydroxy indole, unii-320un7xzyn
16054
1H-indol-5-ol
OC1=CC=C2NC=CC2=C1
This item is not returnable. View return policy
1953-54-4
C8H7NO
133.15
MFCD00005677
LMIQERWZRIFWNZ-UHFFFAOYSA-N
5-hydroxyindole, indol-5-ol, 5-hydroxy-1h-indole, hydroxy-5 indole, 5-hydroxy-indole, 5-indolol, hydroxy-5-indole, 5-hydroxyindol, 5-hydroxy indole, unii-320un7xzyn
16054
1H-indol-5-ol
OC1=CC=C2NC=CC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 105.0°C to 110.0°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Beilstein 21, III, 765
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone,ethanol and acetic acid
Formula Weight 133.15
Percent Purity 97%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 5-Hydroxyindole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Hydroxyindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'