missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-indolecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 10722971
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
21598-06-1
C9H6NO3
176.15
MFCD00005615
BIMHWDJKNOMNLD-UHFFFAOYSA-M
5-hydroxyindole-2-carboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxyindole-2-carboxylicacid, zlchem 754, pubchem1716, acmc-20am33, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxy-1h-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-hydroxy
88958
5-hydroxy-1H-indole-2-carboxylic acid
OC1=CC=C2NC(=CC2=C1)C([O-])=O
This item is not returnable. View return policy
21598-06-1
C9H6NO3
176.15
MFCD00005615
BIMHWDJKNOMNLD-UHFFFAOYSA-M
5-hydroxyindole-2-carboxylic acid, 5-hydroxy-2-indolecarboxylic acid, 5-hydroxyindole-2-carboxylicacid, zlchem 754, pubchem1716, acmc-20am33, 5-hydroxy-indole-2-carboxylic acid, 5-hydroxy-1h-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-hydroxy
88958
5-hydroxy-1H-indole-2-carboxylic acid
OC1=CC=C2NC(=CC2=C1)C([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 249.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 177.16
Percent Purity 98%
Physical Form Fuming Liquid
Chemical Name or Material 5-Hydroxy-2-indolecarboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Hydroxy-2-indolecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'