missing translation for 'onlineSavingsMsg'
Learn More

5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals

Catalog Number 11974621
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
928-90-5
C6H10O
98.15
MFCD00002980
GOQJMMHTSOQIEI-UHFFFAOYSA-N
5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
70234
hex-5-yn-1-ol
OCCCCC#C
This item is not returnable. View return policy
928-90-5
C6H10O
98.15
MFCD00002980
GOQJMMHTSOQIEI-UHFFFAOYSA-N
5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
70234
hex-5-yn-1-ol
OCCCCC#C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 73.0°C to 75.0°C (15.0 mmHg)
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HC≡C(CH2)4OH
Refractive Index 1.4490 to 1.4510
Quantity 25 g
Specific Gravity 0.89
Formula Weight 98.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Hexyn-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • 5-Hexyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'