missing translation for 'onlineSavingsMsg'
Learn More

5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals

Catalog Number p-7058836
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
928-90-5
C6H10O
98.15
MFCD00002980
GOQJMMHTSOQIEI-UHFFFAOYSA-N
5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
70234
hex-5-yn-1-ol
OCCCCC#C
This item is not returnable. View return policy
928-90-5
C6H10O
98.15
MFCD00002980
GOQJMMHTSOQIEI-UHFFFAOYSA-N
5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
70234
hex-5-yn-1-ol
OCCCCC#C

5-Hexyn-1-ol is used as starting material for the preparation of terminal alkynes and lactones containing terminal alykyne groups. It is also used in the synthesis of cinnoline-fused cyclic enediyne, llycopodium alkaloids, (+)-nankakurine A and (+)-nankakurine B and 7-benzoyloxy-3-(2-nitrophenylseleno)-1,5-cyclodecadiyne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 0.880 g/mL
Boiling Point 73°C to 75°C (15 mmHg)
Flash Point 70°C (158°F)
Odor Characteristic
Linear Formula HC≡C(CH2)3CH2OH
Refractive Index 1.4451
Quantity 1 g
Solubility Information Slightly miscible with water.
Formula Weight 98.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Hexyn-1-ol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'