missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-2-one, 98%, ACROS Organics™

£15.38 - £138.81

Chemical Identifiers

CAS 109-49-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.14
MDL Number MFCD00008793
InChI Key RNDVGJZUHCKENF-UHFFFAOYSA-N
Synonym 5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
PubChem CID 7989
IUPAC Name hex-5-en-2-one
SMILES CC(=O)CCC=C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10193381
SDS View Documents
Acros Organics
120780050
5mL Glass bottle £15.38
5mL View alternative products
Save Up to 
»
In stock, ready to ship: 5 Please to purchase this item. Need a web account? Register with us today!
10764241
SDS View Documents
Acros Organics
120780200
20mL Glass bottle £36.37
20mL View alternative products
Save Up to 
»
Estimated Shipment
26-08-2020
Please to purchase this item. Need a web account? Register with us today!
10070970
SDS View Documents
Acros Organics
120781000
100mL Glass bottle £138.81
100mL View alternative products
Save Up to 
»
Estimated Shipment
26-08-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

109-49-9
98.14
RNDVGJZUHCKENF-UHFFFAOYSA-N
7989
CC(=O)CCC=C
C6H10O
MFCD00008793
5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
hex-5-en-2-one

Specifications

5-Hexen-2-one
23°C
1.4190 to 1.421
128°C to 129°C
109-49-9
100.0
C6H10O
MFCD00008793
5-hexen-2-one, allylacetone, 5-hexene-2-one, 1-hexen-5-one, 3-buten-1-yl methyl ketone, unii-xwm6rl9yei, xwm6rl9yei, 1-hexene-5-one, 5-oxo-1-hexene, allyl acetone
RNDVGJZUHCKENF-UHFFFAOYSA-N
hex-5-en-2-one
7989
98%
0.8400g/mL
Glass bottle
0.84
5mL
97.5
97.5% min. (GC)
H2C=CHCH2CH2COCH3
01, 734
Solubility in water: soluble in water. Other solubilities: soluble in organic solvents
CC(=O)CCC=C
98.14
98.14
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Hexen-2-one

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.