missing translation for 'onlineSavingsMsg'
Learn More

5-Hexen-1-ol, 99%, Thermo Scientific Chemicals

Catalog Number 10647162
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
821-41-0
C6H12O
100.16
UIZVMOZAXAMASY-UHFFFAOYSA-N
5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2
69963
hex-5-en-1-ol
C=CCCCCO
This item is not returnable. View return policy
821-41-0
C6H12O
100.16
UIZVMOZAXAMASY-UHFFFAOYSA-N
5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2
69963
hex-5-en-1-ol
C=CCCCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8725g/mL
Boiling Point 152.0°C to 155.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CH(CH2)4OH
Refractive Index 1.4340 to 1.4360
Quantity 5 mL
Beilstein 01,IV,2137
Specific Gravity 0.8725
Solubility Information Solubility in water: 18.6g/L (20°C).
Formula Weight 100.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 5-Hexen-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • 5-Hexen-1-ol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'