missing translation for 'onlineSavingsMsg'
Learn More

5-Formyluracil, 98%

Brand:  Acros Organics 399140010

Code : 29

Additional Details : CAS Number : 1195-08-0 Weight : 0.05100kg


 View more versions of this product

Product Code. 10605125

  • £105.50 / 1g


Estimated Shipment
06-06-2018
Add to basket


Specifications

Chemical Identifiers

5-Formyluracil
1195-08-0
C5H4N2O3
140.1

Specifications

5-Formyluracil
98%
C5H4N2O3
1g
1195-08-0
Glass bottle
140.1
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Formyluracil

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS