missing translation for 'onlineSavingsMsg'
Learn More

5-Formyluracil, 98% , Thermo Scientific Chemicals

Catalog Number p-6827176
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
1195-08-0
C5H4N2O3
140.10
MFCD00192185
OHAMXGZMZZWRCA-UHFFFAOYSA-N
5-formyluracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, uracil 5-carbaldehyde, uracil-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 1,2,3,4-tetrahydro-2,4-dioxo, 2,4 1h,3h-pyrimidinedione, 5-formyl, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxaldehyde, 2,3h-pyrimidinedione, 5-formyl, 5-pyrimidinecarboxaldehyde,2,3,4-tetrahydro-2,4-dioxo, zlchem 870
304590
CHEBI:80961
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde
O=CC1=CNC(=O)NC1=O
This item is not returnable. View return policy
1195-08-0
C5H4N2O3
140.10
MFCD00192185
OHAMXGZMZZWRCA-UHFFFAOYSA-N
5-formyluracil, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, uracil 5-carbaldehyde, uracil-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 1,2,3,4-tetrahydro-2,4-dioxo, 2,4 1h,3h-pyrimidinedione, 5-formyl, 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxaldehyde, 2,3h-pyrimidinedione, 5-formyl, 5-pyrimidinecarboxaldehyde,2,3,4-tetrahydro-2,4-dioxo, zlchem 870
304590
CHEBI:80961
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde
O=CC1=CNC(=O)NC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 98%
Quantity 250 mg
Formula Weight 140.1
Chemical Name or Material 5-Formyluracil
compliance-icons
Product Identifier
  • 5-Formyluracil
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'