missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%, Thermo Scientific Chemicals
42.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 167.16 |
Percent Purity | 96% |
Physical Form | Solid |
Chemical Name or Material | 5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'