missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluorouridine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.
Solubility
Soluble in Methanol (Slightly), Water (Slightly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.
5-Fluorouridine exhibits anticancer activity. It is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs. It is an active metabolite of Doxifluridine.
Solubility
Soluble in Methanol (Slightly), Water (Slightly).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Store at 4°C.
Specifications
Color | White |
Quantity | 5 g |
Beilstein | 33662 |
Solubility Information | Soluble in Methanol (Slightly),Water (Slightly). |
Formula Weight | 262.19 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | 5-Fluorouridine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'