missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoronicotinic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7053606
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
402-66-4
C6H4FNO2
141.10
MFCD01318537
BXZSBDDOYIWMGC-UHFFFAOYSA-N
5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinicacid, pubchem10942
120232
5-fluoropyridine-3-carboxylic acid
OC(=O)C1=CN=CC(F)=C1
This item is not returnable. View return policy
402-66-4
C6H4FNO2
141.10
MFCD01318537
BXZSBDDOYIWMGC-UHFFFAOYSA-N
5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinicacid, pubchem10942
120232
5-fluoropyridine-3-carboxylic acid
OC(=O)C1=CN=CC(F)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoronicotinic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 193°C to 198°C
Color White
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 141.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Fluoronicotinic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'