missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Fluoro-3-(dimethylaminomethyl)indole, 95%

Brand:  Alfa Aesar™ H63918.06

Code : NEW

Additional Details : CAS Number : 343-90-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15400889

  • £478.06 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

5-Fluoro-3-(dimethylaminomethyl)indole
192.23
160256
343-90-8
MFCD00022731
5-Fluorogramine

Specifications

5-Fluoro-3-(dimethylaminomethyl)indole
343-90-8
95%
MFCD00022731
5-Fluorogramine
5g
Solid
192.23
160256
Videos
SDS