missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Fluoro-3-(1-piperazinylmethyl)Indole, 95%

Brand:  Alfa Aesar™ H63645.03

Code : NEW

Additional Details : CAS Number : 147595-45-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15480439

  • £129.60 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Fluoro-3-(1-piperazinylmethyl)Indole
147595-45-7
233.28

Specifications

5-Fluoro-3-(1-piperazinylmethyl)Indole
147595-45-7
95%
1g
Solid
233.28
Videos
SDS