missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-(trifluoromethyl)benzonitrile, 97+%, Thermo Scientific™

Catalog Number p-7027272
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
240800-45-7
C8H3F4N
189.11
MFCD00236281
SQAZPEKSXOYPDX-UHFFFAOYSA-N
5-fluoro-2-trifluoromethyl benzonitrile, 2-cyano-4-fluorobenzotrifluoride, 5-fluoro-2-trifluromethylbenzonitrile, benzonitrile, 5-fluoro-2-trifluoromethyl, 5-fluoro-2-trifluoromethyl benzenecarbonitrile, pubchem14791, ksc495g0j, 5-fluoro-2-trifluoromethyl-benzonitrile, 3-cyano-4-trifluoromethyl-1-fluorobenzene
2737551
5-fluoro-2-(trifluoromethyl)benzonitrile
FC1=CC=C(C(=C1)C#N)C(F)(F)F
This item is not returnable. View return policy
240800-45-7
C8H3F4N
189.11
MFCD00236281
SQAZPEKSXOYPDX-UHFFFAOYSA-N
5-fluoro-2-trifluoromethyl benzonitrile, 2-cyano-4-fluorobenzotrifluoride, 5-fluoro-2-trifluromethylbenzonitrile, benzonitrile, 5-fluoro-2-trifluoromethyl, 5-fluoro-2-trifluoromethyl benzenecarbonitrile, pubchem14791, ksc495g0j, 5-fluoro-2-trifluoromethyl-benzonitrile, 3-cyano-4-trifluoromethyl-1-fluorobenzene
2737551
5-fluoro-2-(trifluoromethyl)benzonitrile
FC1=CC=C(C(=C1)C#N)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3276
Formula Weight 189.11
Percent Purity ≥97%
Chemical Name or Material 5-Fluoro-2-(trifluoromethyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'