missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Fluoro-2-(phenoxycarbonylamino)benzeneboronic acid pinacol ester, 96%

Brand:  Alfa Aesar™ H62984.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478066

  • £352.70 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

5-Fluoro-2-(phenoxycarbonylamino)benzeneboronic acid pinacol ester
357.18
5-Fluoro-2-(phenoxycarbonylamino)phenylboronic acid pinacol ester

Specifications

5-Fluoro-2-(phenoxycarbonylamino)benzeneboronic acid pinacol ester
Solid
357.18
1g
96%
5-Fluoro-2-(phenoxycarbonylamino)phenylboronic acid pinacol ester
Videos
SDS