5-Fluoro-2-nitrobenzoic acid, 98% , ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 398870050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 320-98-9 Weight: 0.05500kg


Packaging Glass bottle
Quantity 5g

Product Code. 10614175

Quantity Price
1 £ 68.2 / 5g
EU Stock 1
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 5-Fluoro-2-nitrobenzoic acid
CAS 320-98-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4FNO4
Formula Weight 185.11
MDL Number MFCD00055635

Specification

Formula Weight 185.11
Physical Form Crystalline Powder
Assay ≥97.5%
Quantity 5g
Color White to pale yellow
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 320-98-9
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00055635
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Molecular Formula C7H4FNO4
CAS Max % 100.0
Chemical Name or Material 5-Fluoro-2-nitrobenzoic acid
Melting Point 130° to 134°C
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.