missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylphenylboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 430650010

Code : 29

Additional Details : CAS Number : 163517-62-2 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 10534238

  • £32.40 / 1g


Estimated Shipment
19-07-2018
Add to basket


Specifications

Chemical Identifiers

5-Fluoro-2-methylphenylboronic acid
96.0
97%
C7H8BFO2
MFCD03095047
163517-62-2
100.0
97%
153.95

Specifications

5-Fluoro-2-methylphenylboronic acid
116.0° to 118.0°C
163517-62-2
100.0
Crystalline Powder
97%
153.95
White
1g
96.0
Authentic
97%
C7H8BFO2
MFCD03095047
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Fluoro-2-methylphenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS